Assay Details
| Gene Symbol: Adamtsl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865942.3 | XP_004865999.1 | 9 - 10 | 1274 | 61 | |
| XM_021245030.1 | XP_021100689.1 | 9 - 10 | 1320 | 61 | ||
| XM_021245031.1 | XP_021100690.1 | 9 - 10 | 1264 | 61 | ||
| XM_021245032.1 | XP_021100691.1 | 9 - 10 | 1217 | 61 | ||
| XM_021245033.1 | XP_021100692.1 | 9 - 10 | 1310 | 61 | ||
| XM_021245034.1 | XP_021100693.1 | 9 - 10 | 1207 | 61 | ||
| XM_021245035.1 | XP_021100694.1 | 9 - 10 | 1274 | 61 | ||
More Information
Set Membership: |
|

Adamtsl5
Genomic Map