Assay Details
| Gene Symbol: Mbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869990.3 | XP_004870047.1 | 8 - 9 | 4620 | 65 | |
| XM_013067524.2 | XP_012922978.1 | 8 - 9 | 3921 | 65 | ||
| XM_021248421.1 | XP_021104080.1 | 8 - 9 | 4515 | 65 | ||
| XM_021248422.1 | XP_021104081.1 | 8 - 9 | 3921 | 65 | ||
More Information
Set Membership: |
|

Mbd5
Genomic Map