Assay Details
| Gene Symbol: Cenpv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870767.3 | XP_004870824.2 | 1 - 2 | 663 | 83 | |
| XM_021248987.1 | XP_021104646.1 | 1 - 2 | 676 | 83 | ||
More Information
Set Membership: |
|

Cenpv
Genomic Map