Assay Details
| Gene Symbol: Cenpv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870767.3 | XP_004870824.2 | 3 - 4 | 840 | 83 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpv| Gene Symbol: Cenpv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870767.3 | XP_004870824.2 | 3 - 4 | 840 | 83 | |