Assay Details
| Gene Symbol: Ctcfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013070763.2 | XP_012926217.2 | 6 - 7 | 1286 | 56 | |
| XM_021255155.1 | XP_021110814.1 | 6 - 7 | 1288 | 56 | ||
More Information
Set Membership: |
|

Ctcfl
Genomic Map