Assay Details
| Gene Symbol: LOC101707673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078974.1 | XP_012934428.1 | 2 - 3 | 98 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101707673| Gene Symbol: LOC101707673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078974.1 | XP_012934428.1 | 2 - 3 | 98 | 63 | |
Set Membership: |
|