Assay Details
| Gene Symbol: LOC101707806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021244890.1 | XP_021100549.1 | 2 - 3 | 421 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101707806| Gene Symbol: LOC101707806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021244890.1 | XP_021100549.1 | 2 - 3 | 421 | 58 | |
Set Membership: |
|