Assay Details
| Gene Symbol: Mpig6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260099.1 | XP_021115758.1 | 4 - 5 | 2712 | 74 | |
| XM_021260100.1 | XP_021115759.1 | 4 - 5 | 2722 | 74 | ||
| XM_021260102.1 | XP_021115761.1 | 4 - 5 | 2763 | 74 | ||
| XM_021260103.1 | XP_021115762.1 | 4 - 5 | 2768 | 74 | ||
| XM_021260104.1 | XP_021115763.1 | 4 - 5 | 829 | 74 | ||
| XM_021260105.1 | XP_021115764.1 | 3 - 4 | 2957 | 74 | ||
More Information
Set Membership: |
|

Mpig6b
Genomic Map