Assay Details
| Gene Symbol: Mpig6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260099.1 | XP_021115758.1 | 5 - 6 | 2744 | 84 | |
| XM_021260100.1 | XP_021115759.1 | 5 - 6 | 2754 | 84 | ||
| XM_021260102.1 | XP_021115761.1 | 5 - 6 | 2795 | 84 | ||
| XM_021260103.1 | XP_021115762.1 | 5 - 6 | 2800 | 84 | ||
| XM_021260104.1 | XP_021115763.1 | 5 - 6 | 861 | 84 | ||
| XM_021260105.1 | XP_021115764.1 | 4 - 5 | 2989 | 84 | ||
More Information
Set Membership: |
|

Mpig6b
Genomic Map