Assay Details
| Gene Symbol: Ncr3lg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263863.1 | XP_021119522.1 | 3 - 4 | 1048 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncr3lg1| Gene Symbol: Ncr3lg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263863.1 | XP_021119522.1 | 3 - 4 | 1048 | 93 | |
Set Membership: |
|