Assay Details
| Gene Symbol: Gpr156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835653.2 | XP_004835710.1 | 5 - 6 | 1146 | 72 | |
| XM_021263056.1 | XP_021118715.1 | 5 - 6 | 1150 | 72 | ||
| XM_021263061.1 | XP_021118720.1 | 5 - 6 | 1146 | 72 | ||
More Information
Set Membership: |
|

Gpr156
Genomic Map