Assay Details
| Gene Symbol: Marveld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838613.2 | XP_004838670.1 | 1 - 2 | 2245 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Marveld1| Gene Symbol: Marveld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838613.2 | XP_004838670.1 | 1 - 2 | 2245 | 60 | |
Set Membership: |
|