Assay Details
Gene Symbol: LOC101698338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004844514.2 | XP_004844571.1 | 24 - 25 | 8195 | 79 | |
XM_004844516.2 | XP_004844573.1 | 24 - 25 | 8192 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
