Assay Details
| Gene Symbol: Glp1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846646.2 | XP_004846703.1 | 9 - 10 | 996 | 60 | |
| XM_021259880.1 | XP_021115539.1 | 9 - 10 | 4046 | 60 | ||
More Information
Set Membership: |
|

Glp1r
Genomic Map