Assay Details
| Gene Symbol: Mrps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872829.2 | XP_004872886.1 | 2 - 3 | 215 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps36| Gene Symbol: Mrps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872829.2 | XP_004872886.1 | 2 - 3 | 215 | 70 | |
Set Membership: |
|