Assay Details
| Gene Symbol: Cep19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834706.3 | XP_004834763.1 | 2 - 3 | 412 | 85 | |
| XM_004834707.3 | XP_004834764.1 | 2 - 3 | 409 | 85 | ||
| XM_004834708.3 | XP_004834765.1 | 2 - 3 | 470 | 85 | ||
| XM_004834709.3 | XP_004834766.1 | 2 - 3 | 467 | 85 | ||
| XM_004834710.3 | XP_004834767.1 | 2 - 3 | 343 | 85 | ||
| XM_004834711.3 | XP_004834768.1 | 2 - 3 | 401 | 85 | ||
More Information
Set Membership: |
|

Cep19
Genomic Map