Assay Details
| Gene Symbol: LOC101711815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834986.3 | XP_004835043.1 | 2 - 3 | 515 | 98 | |
| XM_004834990.3 | XP_004835047.1 | 3 - 4 | 497 | 98 | ||
| XM_004834991.3 | XP_004835048.1 | 3 - 4 | 314 | 98 | ||
| XM_021256964.1 | XP_021112623.1 | 3 - 4 | 614 | 98 | ||
| XM_021256970.1 | XP_021112629.1 | 2 - 3 | 398 | 98 | ||
More Information
Set Membership: |
|

LOC101711815
Genomic Map