Assay Details
| Gene Symbol: LOC101711815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834986.3 | XP_004835043.1 | 4 - 5 | 848 | 92 | |
| XM_004834990.3 | XP_004835047.1 | 5 - 6 | 830 | 92 | ||
| XM_004834991.3 | XP_004835048.1 | 5 - 6 | 647 | 92 | ||
| XM_021256964.1 | XP_021112623.1 | 5 - 6 | 947 | 92 | ||
| XM_021256970.1 | XP_021112629.1 | 4 - 5 | 731 | 92 | ||
More Information
Set Membership: |
|

LOC101711815
Genomic Map