Assay Details
Gene Symbol: CUNH1orf131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004854658.3 | XP_004854715.1 | 4 - 5 | 660 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CUNH1orf131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004854658.3 | XP_004854715.1 | 4 - 5 | 660 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() |