Assay Details
| Gene Symbol: Chrnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857418.3 | XP_004857475.1 | 9 - 10 | 1327 | 63 | |
| XM_004857419.3 | XP_004857476.1 | 9 - 10 | 1327 | 63 | ||
| XM_004857420.3 | XP_004857477.1 | 9 - 10 | 1180 | 63 | ||
| XM_004857421.3 | XP_004857478.1 | 8 - 9 | 1207 | 63 | ||
More Information
Set Membership: |
|

Chrnb1
Genomic Map