Assay Details
| Gene Symbol: Hsd17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859336.2 | XP_004859393.1 | 2 - 3 | 814 | 66 | |
| XM_004859337.2 | XP_004859394.1 | 3 - 4 | 702 | 66 | ||
| XM_021240070.1 | XP_021095729.1 | 2 - 3 | 836 | 66 | ||
| XM_021240071.1 | XP_021095730.1 | 3 - 4 | 730 | 66 | ||
| XM_021240072.1 | XP_021095731.1 | 2 - 3 | 534 | 66 | ||
More Information
Set Membership: |
|

Hsd17b1
Genomic Map