Assay Details
| Gene Symbol: Gsto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866504.3 | XP_004866561.1 | 1 - 2 | 177 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsto1| Gene Symbol: Gsto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866504.3 | XP_004866561.1 | 1 - 2 | 177 | 63 | |
Set Membership: |
|