Assay Details
| Gene Symbol: Il4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866857.3 | XP_004866914.1 | 7 - 8 | 1054 | 74 | |
| XM_004866858.2 | XP_004866915.1 | 6 - 7 | 787 | 74 | ||
More Information
Set Membership: |
|

Il4i1
Genomic Map