Assay Details
| Gene Symbol: Nars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869687.2 | XP_004869744.1 | 11 - 12 | 1463 | 78 | |
| XM_004869688.2 | XP_004869745.1 | 11 - 12 | 1460 | 78 | ||
| XM_004869689.2 | XP_004869746.1 | 10 - 11 | 1382 | 78 | ||
| XM_021248245.1 | XP_021103904.1 | 11 - 12 | 1463 | 78 | ||
More Information
Set Membership: |
|

Nars2
Genomic Map