Assay Details
| Gene Symbol: Oca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872212.3 | XP_004872269.2 | 8 - 9 | 1229 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oca2| Gene Symbol: Oca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872212.3 | XP_004872269.2 | 8 - 9 | 1229 | 55 | |
Set Membership: |
|