Assay Details
| Gene Symbol: Mrps18c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872333.2 | XP_004872390.1 | 2 - 3 | 328 | 118 | |
| XM_021250287.1 | XP_021105946.1 | 4 - 5 | 431 | 118 | ||
More Information
Set Membership: |
|

Mrps18c
Genomic Map