Assay Details
| Gene Symbol: LOC101722487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069651.2 | XP_012925105.1 | 1 | 419 | 100 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101722487| Gene Symbol: LOC101722487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069651.2 | XP_012925105.1 | 1 | 419 | 100 | |
Set Membership: | 
		   
					
						
							 |