Assay Details
| Gene Symbol: Rccd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075089.2 | XP_012930543.1 | 2 - 3 | 894 | 59 | |
| XM_013075090.2 | XP_012930544.1 | 2 - 3 | 903 | 59 | ||
| XM_013075093.2 | XP_012930547.1 | 2 - 3 | 900 | 59 | ||
| XM_021264830.1 | XP_021120489.1 | 2 - 3 | 902 | 59 | ||
More Information
Set Membership: |
|

Rccd1
Genomic Map