Assay Details
| Gene Symbol: LOC101712717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245558.1 | XP_021101217.1 | 3 - 4 | 728 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101712717| Gene Symbol: LOC101712717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245558.1 | XP_021101217.1 | 3 - 4 | 728 | 85 | |
Set Membership: |
|