Assay Details
| Gene Symbol: LOC101709959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247819.1 | XP_021103478.1 | 2 - 3 | 843 | 85 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101709959| Gene Symbol: LOC101709959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247819.1 | XP_021103478.1 | 2 - 3 | 843 | 85 | |