Assay Details
Gene Symbol: Nrarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021261604.1 | XP_021117263.1 | 1 | 320 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
