Assay Details
| Gene Symbol: Ldlrad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265303.1 | XP_021120962.1 | 7 - 8 | 2240 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ldlrad4| Gene Symbol: Ldlrad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265303.1 | XP_021120962.1 | 7 - 8 | 2240 | 55 | |
Set Membership: |
|