Assay Details
| Gene Symbol: Hey2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845937.2 | XP_004845994.1 | 1 - 2 | 196 | 73 | |
| XM_021259214.1 | XP_021114873.1 | 1 - 2 | 196 | 73 | ||
| XM_021259215.1 | XP_021114874.1 | 1 - 2 | 196 | 73 | ||
| XM_021259216.1 | XP_021114875.1 | 1 - 2 | 196 | 73 | ||
More Information
Set Membership: |
|

Hey2
Genomic Map