Assay Details
| Gene Symbol: Cxcl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848038.2 | XP_004848095.1 | 3 - 4 | 434 | 86 | |
| XM_004848039.2 | XP_004848096.1 | 2 - 3 | 323 | 86 | ||
More Information
Set Membership: |
|

Cxcl9
Genomic Map