Assay Details
| Gene Symbol: Kcns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850155.2 | XP_004850212.1 | 1 - 2 | 65 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004850155.2 transcripts.
Set Membership: |
|

Kcns2
Genomic Map