Assay Details
| Gene Symbol: Szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850368.2 | XP_004850425.1 | 3 - 4 | 401 | 60 | |
| XM_004850369.2 | XP_004850426.1 | 3 - 4 | 398 | 60 | ||
| XM_021262869.1 | XP_021118528.1 | 3 - 4 | 333 | 60 | ||
More Information
Set Membership: |
|

Szrd1
Genomic Map