Assay Details
| Gene Symbol: Impa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853257.2 | XP_004853314.1 | 2 - 3 | 267 | 79 | |
| XM_021265268.1 | XP_021120927.1 | 2 - 3 | 273 | 79 | ||
More Information
Set Membership: |
|

Impa2
Genomic Map