Assay Details
| Gene Symbol: Zmym5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854891.3 | XP_004854948.1 | 4 - 5 | 978 | 106 | |
| XM_004854892.3 | XP_004854949.1 | 5 - 6 | 962 | 106 | ||
| XM_021236380.1 | XP_021092039.1 | 5 - 6 | 1091 | 106 | ||
| XM_021236381.1 | XP_021092040.1 | 5 - 6 | 1031 | 106 | ||
| XM_021236382.1 | XP_021092041.1 | 5 - 6 | 961 | 106 | ||
| XM_021236383.1 | XP_021092042.1 | 4 - 5 | 1022 | 106 | ||
| XM_021236384.1 | XP_021092043.1 | 4 - 5 | 967 | 106 | ||
| XM_021236385.1 | XP_021092044.1 | 4 - 5 | 894 | 106 | ||
More Information
Set Membership: |
|

Zmym5
Genomic Map