Assay Details
| Gene Symbol: Tpgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855234.3 | XP_004855291.1 | 3 - 4 | 364 | 69 | |
| XM_004855235.3 | XP_004855292.1 | 3 - 4 | 298 | 69 | ||
| XM_004855237.3 | XP_004855294.1 | 2 - 3 | 239 | 69 | ||
| XM_021236696.1 | XP_021092355.1 | 5 - 6 | 604 | 69 | ||
| XM_021236697.1 | XP_021092356.1 | 5 - 6 | 465 | 69 | ||
| XM_021236698.1 | XP_021092357.1 | 4 - 5 | 471 | 69 | ||
More Information
Set Membership: |
|

Tpgs2
Genomic Map