Assay Details
| Gene Symbol: Dkk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855446.2 | XP_004855503.1 | 1 - 2 | 378 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dkk4| Gene Symbol: Dkk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855446.2 | XP_004855503.1 | 1 - 2 | 378 | 73 | |