Assay Details
Gene Symbol: Dkk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855446.2 | XP_004855503.1 | 2 - 3 | 517 | 70 | |
XM_021236747.1 | XP_021092406.1 | 3 - 4 | 6083 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
