Assay Details
| Gene Symbol: Fgf16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867486.2 | XP_004867543.1 | 2 - 3 | 659 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf16| Gene Symbol: Fgf16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867486.2 | XP_004867543.1 | 2 - 3 | 659 | 103 | |
Set Membership: |
|