Assay Details
| Gene Symbol: Rpp25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872040.3 | XP_004872097.1 | 1 | 919 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004872040.3 transcripts.
Set Membership: |
|

Rpp25
Genomic Map