Assay Details
| Gene Symbol: Nectin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873137.3 | XP_004873194.1 | 6 - 7 | 1426 | 112 | |
| XM_021250954.1 | XP_021106613.1 | 6 - 7 | 1427 | 112 | ||
More Information
Set Membership: |
|

Nectin2
Genomic Map