Assay Details
| Gene Symbol: LOC101717069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066515.2 | XP_012921969.1 | 20 - 21 | 1920 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101717069| Gene Symbol: LOC101717069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066515.2 | XP_012921969.1 | 20 - 21 | 1920 | 112 | |
Set Membership: |
|