Assay Details
| Gene Symbol: Mmp17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257695.1 | XP_021113354.1 | 4 - 5 | 757 | 61 | |
| XM_021257696.1 | XP_021113355.1 | 4 - 5 | 757 | 61 | ||
| XM_021257697.1 | XP_021113356.1 | 4 - 5 | 766 | 61 | ||
More Information
Set Membership: |
|

Mmp17
Genomic Map