Assay Details
Gene Symbol: Skil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834583.3 | XP_004834640.1 | 3 - 4 | 2099 | 83 | |
XM_021254089.1 | XP_021109748.1 | 3 - 4 | 1955 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
