Assay Details
| Gene Symbol: Rhpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837842.3 | XP_004837899.1 | 11 - 12 | 1578 | 71 | |
| XM_004837843.3 | XP_004837900.1 | 11 - 12 | 1580 | 71 | ||
| XM_013069486.2 | XP_012924940.1 | 10 - 11 | 1358 | 71 | ||
More Information
Set Membership: |
|

Rhpn1
Genomic Map