Assay Details
| Gene Symbol: LOC101726570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838016.3 | XP_004838073.1 | 5 - 6 | 806 | 87 | |
| XM_004838018.3 | XP_004838075.1 | 4 - 5 | 715 | 87 | ||
| XM_021252518.1 | XP_021108177.1 | 5 - 6 | 602 | 87 | ||
More Information
Set Membership: |
|

LOC101726570
Genomic Map