Assay Details
| Gene Symbol: LOC101697932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839947.3 | XP_004840004.1 | 2 - 3 | 425 | 101 | |
| XM_004839948.3 | XP_004840005.1 | 2 - 3 | 357 | 101 | ||
| XM_004839953.3 | XP_004840010.1 | 2 - 3 | 425 | 101 | ||
| XM_021254297.1 | XP_021109956.1 | 2 - 3 | 565 | 101 | ||
| XM_021254298.1 | XP_021109957.1 | 2 - 3 | 497 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004839947.3, XM_004839948.3, XM_004839953.3, XM_021254297.1, XM_021254298.1 transcripts.
Set Membership: |
|

LOC101697932
Genomic Map